siRNA Details
| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145151.3 | NP_660133.2 | 1 | 1291 | ||
| GenBank mRNA | AA028814.1 | - | 4 | 327 | ||
| AF408398.1 | - | 1 | 963 | |||
| BG094109.1 | - | 1 | 403 | |||
| BI990274.1 | - | 1 | 230 | |||
| CX237465.1 | - | 1 | 343 | |||

Crebzf
Genomic Map