siRNA Details
| Gene Symbol: Plekha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040271.1 | - | 10 | 1758 | ||
| AK048843.1 | - | 10 | 1924 | |||
| AK146866.1 | - | 1949 | ||||
| AK160810.1 | - | 10 | 1838 | |||
| BC060678.1 | - | 9 | 1679 | |||
| BC084587.1 | - | 10 | 1853 | 3602481 | ||
| EU380771.1 | - | 1975 | ||||
| EU380772.1 | - | 2033 | ||||

Plekha7
Genomic Map