siRNA Details
| Gene Symbol: Plekha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040271.1 | - | 11 | 1815 | ||
| AK146866.1 | - | 2006 | ||||
| AK160810.1 | - | 11 | 1895 | |||
| BC060678.1 | - | 10 | 1736 | |||
| BC084587.1 | - | 11 | 1910 | 3602481 | ||
| EU380771.1 | - | 2032 | ||||
| EU380772.1 | - | 2090 | ||||

Plekha7
Genomic Map