siRNA Details
| Gene Symbol: Xylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175645.3 | NP_783576.2 | 12 | 3027 | ||
| GenBank mRNA | AJ539164.1 | - | 12 | 3029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xylt1| Gene Symbol: Xylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175645.3 | NP_783576.2 | 12 | 3027 | ||
| GenBank mRNA | AJ539164.1 | - | 12 | 3029 | ||