siRNA Details
| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177697.3 | NP_808365.2 | 14, 15 | 1732 | ||
| GenBank mRNA | AK086942.1 | - | 13, 14 | 1330 | ||
| AK132732.1 | - | 14, 15 | 1733 | |||
| AK136898.1 | - | 13, 14 | 1331 | |||

Vwa3a
Genomic Map