siRNA Details
| Gene Symbol: Zfp764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167832.1 | NP_001161304.1 | 3 | 972 | ||
| NM_146203.4 | NP_666315.3 | 3 | 1009 | |||
| GenBank mRNA | AK033218.1 | - | 3 | 1001 | ||
| AK042873.1 | - | 3 | 1010 | |||
| AK053795.1 | - | 3 | 934 | |||
| BC026397.1 | - | 3 | 894 | 4207307 | ||

Zfp764
Genomic Map