siRNA Details
| Gene Symbol: Prr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145589.2 | NP_663564.2 | 3 | 606 | ||
| GenBank mRNA | AK133884.1 | - | 3 | 571 | ||
| AK139559.1 | - | 4 | 381 | |||
| AK145816.1 | - | 4 | 386 | |||
| AK151203.1 | - | 4 | 438 | |||
| BC055033.1 | - | 3 | 606 | 6310272 | ||

Prr14
Genomic Map