siRNA Details
| Gene Symbol: Tpcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146206.4 | NP_666318.2 | 21 | 1829 | ||
| GenBank mRNA | AK052930.1 | - | 21 | 1830 | ||
| AK089490.1 | - | 21 | 1893 | |||
| AK090059.1 | - | 19 | 1654 | |||
| AK170863.1 | - | 20 | 1775 | |||
| BC025890.1 | - | 4 | 713 | |||
| BC043472.1 | - | 17 | 1407 | |||
| BC069857.1 | - | 4 | 517 | |||
| BC141195.1 | - | 21 | 1913 | 9055807 | ||

Tpcn2
Genomic Map