siRNA Details
| Gene Symbol: Zfp958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145591.4 | NP_663566.1 | 4 | 1378 | ||
| GenBank mRNA | AK037083.1 | - | 4 | 1379 | ||
| AK037596.1 | - | 4 | 1381 | |||
| BC003267.1 | - | 4 | 1381 | 3499094 | ||
| BC138223.1 | - | 4 | 1325 | 8861242 | ||
| BC145996.1 | - | 4 | 1325 | 40131299 | ||

Zfp958
Genomic Map