siRNA Details
| Gene Symbol: Erich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034862.2 | NP_001030034.2 | 5 | 839 | ||
| GenBank mRNA | AK147828.1 | - | 5 | 814 | ||
| AK170294.1 | - | 3 | 504 | |||
| CF981481.1 | - | 2 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich1| Gene Symbol: Erich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034862.2 | NP_001030034.2 | 5 | 839 | ||
| GenBank mRNA | AK147828.1 | - | 5 | 814 | ||
| AK170294.1 | - | 3 | 504 | |||
| CF981481.1 | - | 2 | 416 | |||