siRNA Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156746.1 | - | 2 | 678 | ||
AK170040.1 | - | 2 | 741 | |||
AK172238.1 | - | 2 | 704 | |||
BC036336.1 | - | 1 | 270 | |||
BC064447.1 | - | 2 | 716 |
