siRNA Details
| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145595.2 | NP_663570.2 | 4, 5 | 651 | ||
| GenBank mRNA | AK031756.1 | - | 3, 4 | 2093 | ||
| AK150763.1 | - | 4, 5 | 652 | |||
| BC009118.1 | - | 4, 5 | 601 | 3154595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbr4| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145595.2 | NP_663570.2 | 4, 5 | 651 | ||
| GenBank mRNA | AK031756.1 | - | 3, 4 | 2093 | ||
| AK150763.1 | - | 4, 5 | 652 | |||
| BC009118.1 | - | 4, 5 | 601 | 3154595 | ||