siRNA Details
| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145595.2 | NP_663570.2 | 5 | 778 | ||
| GenBank mRNA | AK031756.1 | - | 4 | 2220 | ||
| AK043313.1 | - | 5 | 603 | |||
| AK150763.1 | - | 5 | 779 | |||
| BC009118.1 | - | 5 | 728 | 3154595 | ||

Cbr4
Genomic Map