siRNA Details
| Gene Symbol: Trim60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153097.2 | NP_694737.1 | 2 | 523 | ||
| GenBank mRNA | AF290197.1 | - | 2 | 425 | ||
| AK135841.1 | - | 2 | 420 | |||
| BC139468.1 | - | 2 | 407 | 8862490 | ||
| BC139469.1 | - | 2 | 407 | 8862491 | ||

Trim60
Genomic Map