siRNA Details
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252623.1 | NP_001239552.1 | 10 | 2297 | ||
NM_172753.5 | NP_766341.4 | 9 | 1993 | |||
GenBank mRNA | AK028688.1 | - | 9 | 1916 | ||
AK033522.1 | - | 7 | 1647 | |||
AK080687.1 | - | 9 | 1962 | |||
AK133567.1 | - | 7 | 1646 | |||
AK144089.1 | - | 6 | 1554 | |||
BC026599.1 | - | 4 | 413 | |||
BC057630.1 | - | 11 | 2265 | |||
BC089162.1 | - | 9 | 1990 | 30470205 |
