siRNA Details
| Gene Symbol: Ccdc102a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033533.3 | NP_001028705.2 | 8 | 1752 | ||
| GenBank mRNA | AK144530.1 | - | 8 | 1709 | ||
| AK165668.1 | - | 8 | 1709 | |||
| BC027663.1 | - | 419 | ||||
| BC107374.2 | - | 7 | 1209 | |||
| BC107375.1 | - | 7 | 1212 | |||
| BC114992.1 | - | 7 | 1214 | |||
| BC139307.1 | - | 7 | 1597 | 8862329 | ||
| BC139308.1 | - | 7 | 1597 | 8862330 | ||

Ccdc102a
Genomic Map