siRNA Details
Gene Symbol: Il17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145834.3 | NP_665833.3 | 1 | 148 | ||
GenBank mRNA | AF458061.1 | - | 1 | 148 | ||
BC145853.1 | - | 1 | 211 | 40131156 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145834.3 | NP_665833.3 | 1 | 148 | ||
GenBank mRNA | AF458061.1 | - | 1 | 148 | ||
BC145853.1 | - | 1 | 211 | 40131156 |