siRNA Details
| Gene Symbol: Chmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145606.3 | NP_663581.1 | 5 | 422 | ||
| GenBank mRNA | AK046416.1 | - | 5 | 395 | ||
| AK159830.1 | - | 5 | 395 | |||
| BC010524.1 | - | 5 | 372 | 3500937 | ||
| BC023807.1 | - | 5 | 385 | 5354266 | ||
| BC036138.1 | - | 5 | 374 | 4972627 | ||
| BC036152.1 | - | 5 | 376 | 4977509 | ||

Chmp1a
Genomic Map