siRNA Details
| Gene Symbol: Med17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144933.1 | NP_659182.1 | 2 | 358 | ||
| GenBank mRNA | AK033151.1 | - | 2 | 364 | ||
| AK038451.1 | - | 2 | 398 | |||
| AK049112.1 | - | 2 | 398 | |||
| AK142033.1 | - | 2 | 397 | |||
| AK151377.1 | - | 2 | 398 | |||
| AK193003.1 | - | 2 | 107 | |||
| BC004062.1 | - | 1 | 35 | |||
| BC020185.1 | - | 2 | 358 | 4505959 | ||

Med17
Genomic Map