siRNA Details
| Gene Symbol: Ppan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145610.2 | NP_663585.2 | 11 | 1398 | ||
| GenBank mRNA | AK039027.1 | - | 11 | 1303 | ||
| AK085695.1 | - | 1 | 3642 | |||
| AK171672.1 | - | 7 | 1064 | |||
| AK172615.1 | - | 7 | 1064 | |||
| BB749945.1 | - | 2 | 163 | |||
| BC014688.1 | - | 11 | 1316 | 4489442 | ||
| BC059277.1 | - | 9 | 1562 | |||

Ppan
Genomic Map