siRNA Details
| Gene Symbol: Zfp809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172763.3 | NP_766351.3 | 6 | 1282 | ||
| GenBank mRNA | BC099418.1 | - | 6 | 1214 | 30525533 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp809| Gene Symbol: Zfp809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172763.3 | NP_766351.3 | 6 | 1282 | ||
| GenBank mRNA | BC099418.1 | - | 6 | 1214 | 30525533 | |