siRNA Details
| Gene Symbol: Adamts15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024139.1 | NP_001019310.1 | 8 | 2387 | ||
| GenBank mRNA | BC009667.1 | - | 3 | 343 | ||
| BC043308.1 | - | 6 | 1008 | |||
| BC094677.1 | - | 8 | 2387 | 30619053 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts15| Gene Symbol: Adamts15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024139.1 | NP_001019310.1 | 8 | 2387 | ||
| GenBank mRNA | BC009667.1 | - | 3 | 343 | ||
| BC043308.1 | - | 6 | 1008 | |||
| BC094677.1 | - | 8 | 2387 | 30619053 | ||