siRNA Details
| Gene Symbol: Foxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291448.1 | NP_001278377.1 | 11 | 1450 | ||
| NM_001291449.1 | NP_001278378.1 | 11 | 1455 | |||
| NM_172291.2 | NP_758495.1 | 11 | 1468 | |||
| GenBank mRNA | AK144759.1 | - | 11 | 1394 | ||
| AK163722.1 | - | 11 | 1309 | |||
| BC017604.1 | - | 11 | 1408 | |||
| BC024806.1 | - | 11 | 1338 | 5361053 | ||
| BC027386.1 | - | 8 | 1929 | |||
| BC028892.1 | - | 9 | 2421 | |||

Foxred1
Genomic Map