siRNA Details
| Gene Symbol: Foxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291448.1 | NP_001278377.1 | 6 | 853 | ||
| NM_001291449.1 | NP_001278378.1 | 6 | 858 | |||
| NM_172291.2 | NP_758495.1 | 6 | 853 | |||
| GenBank mRNA | AK054231.1 | - | 6 | 745 | ||
| AK144759.1 | - | 6 | 779 | |||
| AK163722.1 | - | 6 | 712 | |||
| BC017604.1 | - | 6 | 811 | |||
| BC024806.1 | - | 6 | 723 | 5361053 | ||
| BC027386.1 | - | 3 | 1332 | |||
| BC028892.1 | - | 4 | 1824 | |||

Foxred1
Genomic Map