siRNA Details
| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172769.2 | NP_766357.1 | 6 | 794 | ||
| GenBank mRNA | AB016248.1 | - | 6 | 780 | ||
| AK043825.1 | - | 6 | 753 | |||
| AK077670.1 | - | 6 | 752 | |||
| AK154309.1 | - | 6 | 741 | |||
| BC024132.1 | - | 6 | 743 | 5101986 | ||

Sc5d
Genomic Map