siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 10 | 1820 | ||
GenBank mRNA | AK028768.1 | - | 10 | 1455 | ||
AK030208.1 | - | 10 | 1452 | |||
AK032376.1 | - | 10 | 886 | |||
AK033867.1 | - | 10 | 1316 | |||
AK051527.1 | - | 10 | 1821 | |||
AK078706.1 | - | 5 | 354 | |||
AK136229.1 | - | 5 | 345 | |||
BC023073.1 | - | 2 | 98 | |||
BC049909.1 | - | 10 | 961 | |||
BC094247.1 | - | 9 | 1305 | 5717984 |
