siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 3 | 688 | ||
GenBank mRNA | AK028768.1 | - | 3 | 323 | ||
AK030208.1 | - | 3 | 320 | |||
AK033867.1 | - | 3 | 184 | |||
AK046190.1 | - | 3 | 673 | |||
AK046997.1 | - | 313 | ||||
AK051527.1 | - | 3 | 689 | |||
BC094247.1 | - | 2 | 173 | 5717984 |
