siRNA Details
| Gene Symbol: Hykk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177351.4 | NP_796325.2 | 5 | 1640 | ||
| GenBank mRNA | AK048771.1 | - | 4 | 1473 | ||
| AK085527.1 | - | 5 | 1649 | |||
| AK138851.1 | - | 5 | 1626 | |||
| AK162425.1 | - | 5 | 1641 | |||
| BC038618.1 | - | 5 | 1619 | 4221933 | ||

Hykk
Genomic Map