siRNA Details
| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031141.1 | - | 5 | 531 | ||
| AK158516.1 | - | 3 | 359 | |||
| AK161676.1 | - | 5 | 588 | |||
| BC017156.1 | - | 5 | 652 | 4911201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp3| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031141.1 | - | 5 | 531 | ||
| AK158516.1 | - | 3 | 359 | |||
| AK161676.1 | - | 5 | 588 | |||
| BC017156.1 | - | 5 | 652 | 4911201 | ||