siRNA Details
| Gene Symbol: Fam214a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113283.1 | NP_001106754.1 | 6 | 2212 | ||
| NM_153584.2 | NP_705812.2 | 5 | 2035 | |||
| GenBank mRNA | AK173161.1 | - | 6 | 2083 | ||
| BC031353.1 | - | 5 | 2035 | 5363230 | ||
| BC060215.1 | - | 6 | 2210 | |||

Fam214a
Genomic Map