siRNA Details
| Gene Symbol: Ppp2r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161362.3 | NP_001154834.1 | 2 | 1439 | ||
| GenBank mRNA | AK042943.1 | - | 2 | 1403 | ||
| AK076163.1 | - | 1 | 486 | |||
| BC042841.1 | - | 1 | 401 | |||
| BC042842.1 | - | 1 | 874 | |||
| BC151170.1 | - | 1 | 1266 | 9088059 | ||
| BC151171.1 | - | 1 | 1261 | 9088060 | ||

Ppp2r3a
Genomic Map