siRNA Details
| Gene Symbol: Glyctk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039586.1 | NP_001034675.1 | 5 | 931 | ||
| NM_174846.4 | NP_777271.3 | 4 | 1067 | |||
| GenBank mRNA | AF455785.1 | - | 5 | 900 | ||
| AK049425.1 | - | 4 | 1048 | |||
| AK050182.1 | - | 5 | 921 | |||
| AK052709.1 | - | 4 | 1006 | |||
| BC025834.1 | - | 4 | 1036 | 5132437 | ||
| BC025935.1 | - | 1028 | 5097665 | |||
| BC030732.1 | - | 4 | 749 | 5102500 | ||
| BC030736.1 | - | 3 | 316 | |||
| BC033063.1 | - | 4 | 1043 | 5100009 | ||
| BC036136.1 | - | 888 | 4971642 | |||
| Gene Symbol: D030055H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031761.1 | - | 1 | 1065 | ||

Glyctk
Genomic Map