siRNA Details
| Gene Symbol: Gm590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195437.1 | NP_001182366.1 | 3 | 348 | ||
| GenBank mRNA | AI536220.1 | - | 3 | 348 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA123812.1 | - | 348 | Chr. - - [-] on Build | ||

Gm590
Genomic Map