siRNA Details
| Gene Symbol: Phgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016966.3 | NP_058662.2 | 8 | 1045 | ||
| GenBank mRNA | AK076815.1 | - | 8 | 1001 | ||
| AK147946.1 | - | 8 | 994 | |||
| AK169684.1 | - | 8 | 1001 | |||
| AK198805.1 | - | 1 | 28 | |||
| BC086668.1 | - | 8 | 953 | 30633243 | ||
| BC110673.1 | - | 8 | 987 | 6311873 | ||
| L21027.1 | - | 7 | 764 | |||

Phgdh
Genomic Map