siRNA Details
| Gene Symbol: Gbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172777.3 | NP_766365.1 | 5 | 569 | ||
| GenBank mRNA | AK088861.1 | - | 5 | 555 | ||
| AK165231.1 | - | 554 | ||||
| BC057170.1 | - | 558 | 4978410 | |||
| CA535632.1 | - | 5 | 569 | |||
| DQ985742.1 | - | 553 | ||||

Gbp9
Genomic Map