siRNA Details
| Gene Symbol: Smok2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167913.2 | NP_001161385.1 | 2 | 615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smok2b| Gene Symbol: Smok2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167913.2 | NP_001161385.1 | 2 | 615 | ||