siRNA Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145628.4 | NP_663603.3 | 13 | 1786 | ||
| GenBank mRNA | AK031767.1 | - | 13 | 1765 | ||
| AK049053.1 | - | 13 | 2200 | |||
| AK169116.1 | - | 5 | 609 | |||
| AK220478.1 | - | 12 | 1499 | |||
| BC005470.1 | - | 11 | 1355 | 3496607 | ||

Usp11
Genomic Map