siRNA Details
| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172779.4 | NP_766367.1 | 17 | 2939 | ||
| GenBank mRNA | AK083942.1 | - | 17 | 2901 | ||
| AK144026.1 | - | 7 | 2784 | |||
| BC019773.1 | - | 3 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx26b| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172779.4 | NP_766367.1 | 17 | 2939 | ||
| GenBank mRNA | AK083942.1 | - | 17 | 2901 | ||
| AK144026.1 | - | 7 | 2784 | |||
| BC019773.1 | - | 3 | 399 | |||