siRNA Details
| Gene Symbol: Mmgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146234.3 | NP_666346.1 | 1, 2 | 142 | ||
| GenBank mRNA | AK036253.1 | - | 1, 2 | 143 | ||
| AK043524.1 | - | 1, 2 | 156 | |||
| AK049689.1 | - | 1, 2 | 117 | |||
| AK163269.1 | - | 1, 2 | 294 | |||
| BC032271.1 | - | 1, 2 | 168 | 4954726 | ||
| EU069461.1 | - | 1, 2 | 101 | |||

Mmgt1
Genomic Map