siRNA Details
| Gene Symbol: Tbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011603.5 | NP_035733.1 | 5 | 720 | ||
| GenBank mRNA | AB017697.1 | - | 5 | 617 | ||
| AK005604.1 | - | 5 | 540 | |||
| AK049975.1 | - | 5 | 592 | |||
| AK078547.1 | - | 5 | 652 | |||
| BC033926.1 | - | 5 | 561 | 1382180 | ||

Tbpl1
Genomic Map