siRNA Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172787.2 | NP_766375.1 | 20, 21 | 2537 | ||
| GenBank mRNA | AK030412.1 | - | 7, 8 | 1135 | ||
| AK045667.1 | - | 20, 21 | 2538 | |||
| AK129443.1 | - | 16, 17 | 2082 | |||
| BC082309.1 | - | 19, 20 | 2407 | 30356672 | ||
| BC085192.1 | - | 19, 20 | 2642 | 30694487 | ||

L3mbtl3
Genomic Map