siRNA Details
| Gene Symbol: Zfp938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105557.2 | NP_001099027.2 | 2, 3 | 227 | ||
| GenBank mRNA | AK045871.1 | - | 2, 3 | 233 | ||
| BC040349.1 | - | 2, 3 | 233 | 4024367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp938| Gene Symbol: Zfp938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105557.2 | NP_001099027.2 | 2, 3 | 227 | ||
| GenBank mRNA | AK045871.1 | - | 2, 3 | 233 | ||
| BC040349.1 | - | 2, 3 | 233 | 4024367 | ||