siRNA Details
| Gene Symbol: Gm4925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037166.2 | NP_001032243.1 | 1 | 552 | ||
| GenBank mRNA | BC100581.1 | - | 1 | 583 | 6435651 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4925| Gene Symbol: Gm4925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037166.2 | NP_001032243.1 | 1 | 552 | ||
| GenBank mRNA | BC100581.1 | - | 1 | 583 | 6435651 | |