siRNA Details
| Gene Symbol: Ptprq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081432.1 | NP_001074901.1 | 27 | 4839 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprq| Gene Symbol: Ptprq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081432.1 | NP_001074901.1 | 27 | 4839 | ||
Set Membership: |
|