siRNA Details
Gene Symbol: Sowaha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183173.2 | NP_898996.2 | 1 | 468 | ||
GenBank mRNA | AK043538.1 | - | 1 | 353 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sowaha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183173.2 | NP_898996.2 | 1 | 468 | ||
GenBank mRNA | AK043538.1 | - | 1 | 353 |