siRNA Details
| Gene Symbol: Slc13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004148.4 | NP_001004148.1 | 9, 10 | 1334 | ||
| GenBank mRNA | AK048662.1 | - | 9, 10 | 1335 | ||
| AK140943.1 | - | 9, 10 | 1336 | |||
| AK158253.1 | - | 9, 10 | 1339 | |||
| AY368893.1 | - | 9, 10 | 1295 | |||

Slc13a5
Genomic Map