siRNA Details
| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 17 | 2295 | ||
| GenBank mRNA | AK052040.1 | - | 17 | 2320 | ||
| AK129379.1 | - | 13 | 1676 | |||
| AY351699.1 | - | 17 | 2295 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdk2| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 17 | 2295 | ||
| GenBank mRNA | AK052040.1 | - | 17 | 2320 | ||
| AK129379.1 | - | 13 | 1676 | |||
| AY351699.1 | - | 17 | 2295 | |||