siRNA Details
| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001333.2 | NP_001001333.2 | 11, 12 | 1616 | ||
| NM_001146073.1 | NP_001139545.1 | 10, 11 | 1421 | |||
| GenBank mRNA | AK154237.1 | - | 11, 12 | 1590 | ||
| BC069960.1 | - | 10, 11 | 1373 | 5007884 | ||
| FM204885.1 | - | 10, 11 | 1153 | |||
| FM204886.1 | - | 10, 11 | 1372 | |||

Hexdc
Genomic Map