siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011790.4 | NP_035920.1 | 13 | 1495 | ||
| GenBank mRNA | AF124664.1 | - | 12 | 1322 | ||
| AJ130975.1 | - | 12 | 1354 | |||
| AK011094.1 | - | 13 | 1502 | |||
| AK045012.1 | - | 1478 | ||||
| AK149936.1 | - | 13 | 1475 | |||
| AK167104.1 | - | 13 | 1452 | |||
| AK168055.1 | - | 13 | 1498 | |||
| BC051998.1 | - | 13 | 1495 | 6400287 | ||
| BC052422.1 | - | 13 | 1495 | 6400287 | ||

Arih2
Genomic Map