siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011790.4 | NP_035920.1 | 3 | 528 | ||
| GenBank mRNA | AF124664.1 | - | 2 | 355 | ||
| AJ130975.1 | - | 2 | 387 | |||
| AK011094.1 | - | 3 | 532 | |||
| AK045012.1 | - | 511 | ||||
| AK149936.1 | - | 3 | 508 | |||
| AK165028.1 | - | 3 | 466 | |||
| AK167104.1 | - | 3 | 485 | |||
| AK168055.1 | - | 3 | 533 | |||
| BC051998.1 | - | 3 | 528 | 6400287 | ||
| BC052422.1 | - | 3 | 528 | 6400287 | ||

Arih2
Genomic Map